Learn More
Aviva Systems Biology Corp PML Antibody - N-terminal region (ARP38184_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38184_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PML. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PML. Alternate names: MYL, PP8675, RNF71, TRIM19ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56556