Learn More
Aviva Systems Biology Corp PML Antibody - C-terminal region (ARP33048_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33048_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PML. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PML. Alternate names: MYL, PP8675, RNF71, TRIM19ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54861