Learn More
New England Biolabs, Inc. PmlI – 10000 units
SDP
Catalog No. 50812195
Encompass
A restriction endonuclease that recognizes the sequence CAC_^GTG.
Catalog No. 50-812-195 Supplier New England Biolabs, Inc. Supplier No. R0532L
A restriction endonuclease that recognizes the sequence CAC_^GTG.