Learn More
New England Biolabs, Inc. PmlI – 2000 units
SDP
Catalog No. 50812196
Encompass
A restriction endonuclease that recognizes the sequence CAC_^GTG.
Catalog No. 50-812-196 Supplier New England Biolabs, Inc. Supplier No. R0532S
A restriction endonuclease that recognizes the sequence CAC_^GTG.