Learn More
Aviva Systems Biology Corp PMM1 Antibody - N-terminal region (ARP56401_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56401_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PMM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PMM1. Alternate names: Sec53ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61843