Learn More
Aviva Systems Biology Corp PMM2 Antibody - middle region (ARP63938_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63938_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PMM2. It was validated on Western Blot. . Alternate names: CDG1, CDG1a, CDGS, PMM 2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-123