Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Standards Kit, Polymethylmethacrylate, Low MW, GPC, 10 Vials

Catalog No. 50785370 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This kit offers a unique collection of 10 Gel Permeation Chromatography (GPC) standards for non-aqueous molecular weight determination. It provides 10 vials, each containing 0.5 g of polymethylmethacrylate low molecular weight standards, covering a range of molecular weights for precise calibration.

  • 10 vials, 0.5 g in each vial
  • Contains molecular weight standards: 1000, 1700, 2500, 3500, 5000, 7000, 10,000, 13,000, 20,000, 30,000
  • Separation mode: GPC/GFC/SEC
  • Detection method: UV, RI/ELSD
  • Formulation: Solid
  • Storage condition: 4°C (fridge)
  • System type: LC
  • Shelf life: 36 months
  • Shelf life upon reconstitution: 1 week
  • Shipping condition: Ambient
  • Tolerance: 10% (Mp)
  • Type: Calibration

Catalog No. 50-785-370 Supplier Waters Corp Supplier No. WAT035707
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This kit offers a unique collection of 10 Gel Permeation Chromatography (GPC) standards for non-aqueous molecular weight determination. It provides 10 vials, each containing 0.5 g of polymethylmethacrylate low molecular weight standards, covering a range of molecular weights for precise calibration.

  • 10 vials, 0.5 g in each vial
  • Contains molecular weight standards: 1000, 1700, 2500, 3500, 5000, 7000, 10,000, 13,000, 20,000, 30,000
  • Separation mode: GPC/GFC/SEC
  • Detection method: UV, RI/ELSD
  • Formulation: Solid
  • Storage condition: 4°C (fridge)
  • System type: LC
  • Shelf life: 36 months
  • Shelf life upon reconstitution: 1 week
  • Shipping condition: Ambient
  • Tolerance: 10% (Mp)
  • Type: Calibration

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.