Learn More
Cayman Chemical PMMoV RT-qPCR Assay, 1 ea
SDP
Catalog No. 502945459
Encompass_Preferred
A mix of primers and probe targeting targeting PMMoV genomic RNA is provided in a tube (a 20X concentrated working solution). Organism Type: Virus (+)ssRNA Enzyme Master Mixes (recommended): Empirical RT-qPCR mastermix, highQU RT-qPCR mastermix, Standard RT-qPCR mastermix, Taqpath RT-qPCR mastermix Panels: Control, Wastewater Instruments: Any instrument
Catalog No. 50-294-5459 Supplier Cayman Chemical Supplier No. 406811 EA
A mix of primers and probe targeting targeting PMMoV genomic RNA is provided in a tube (a 20X concentrated working solution). Organism Type: Virus (+)ssRNA Enzyme Master Mixes (recommended): Empirical RT-qPCR mastermix, highQU RT-qPCR mastermix, Standard RT-qPCR mastermix, Taqpath RT-qPCR mastermix Panels: Control, Wastewater Instruments: Any instrument