Learn More
Cayman Chemical Pimozide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1803193-57-8, 1 mg
SDP
Catalog No. 502939274
Encompass_Preferred
An internal standard for the quantification of pimozide by GC- or LC-MS
Catalog No. 50-293-9274 Supplier Cayman Chemical Supplier No. 286171 MG
An internal standard for the quantification of pimozide by GC- or LC-MS