Learn More
Biocare Medical, LLC PMS2 0.5ML
SDP
Supplier:  Biocare Medical, LLC CM344BK
Encompass_Preferred
PMS2 (post meiotic segregration increased 2) is one of the four major DNA mismatch repair genes. Studies show that the MSI in endometrial carcinoma is linked to loss of MLH1/PMS2. PMS2 expression may be useful to screen for Lynch syndrome.
Catalog No. 50-826-50