Learn More
Diagnostic Biosystems PMS2 PMS2
Catalog No. 502407465
Encompass_Preferred
Postmeiotic segregation increased 2 or PMS2 was originally discovered in S. cerevisiae and is part of the mismatch repair system. It resides on 7p22.2 and its gene product partners with MLH1 to help detect mismatches in DNA. Mutations in PMS2 have been reported in about two percent of families with Lynch syndrome (hereditary nonpolyposis colorectal cancer).
Catalog No. 50-240-7465 Supplier Diagnostic Biosystems Supplier No. PDM171
Postmeiotic segregation increased 2 or PMS2 was originally discovered in S. cerevisiae and is part of the mismatch repair system. It resides on 7p22.2 and its gene product partners with MLH1 to help detect mismatches in DNA. Mutations in PMS2 have been reported in about two percent of families with Lynch syndrome (hereditary nonpolyposis colorectal cancer).