Learn More
Research Products International Corp PMSF [Phenylmethylsulfonyl fluoride], 1 Gram
SDP
Catalog No. 501368487
Encompass_Preferred
Irreversible serine protease inhibitor of chymotrypsin, kallikrein, papain, subtilisin, thrombin and trypsin. Working Conentration: 0.1-1.0mM.
Catalog No. 50-136-8487 Supplier Research Products International Corp Supplier No. P202701.0
Irreversible serine protease inhibitor of chymotrypsin, kallikrein, papain, subtilisin, thrombin and trypsin. Working Conentration: 0.1-1.0mM.