Learn More
Research Products International Corp PMSF 10 Grams
SDP
Catalog No. 502388445
Encompass_Preferred
Irreversible serine protease inhibitor of chymotrypsin, kallikrein, papain, subtilisin, thrombin and trypsin. Working Conentration: 0.1-1.0mM.
Catalog No. 50-238-8445 Supplier Research Products International Corp Supplier No. P2027010.0
Irreversible serine protease inhibitor of chymotrypsin, kallikrein, papain, subtilisin, thrombin and trypsin. Working Conentration: 0.1-1.0mM.