Learn More
Research Products International Corp PMSF [Phenylmethylsulfonyl fluoride], 25 Grams
SDP
Catalog No. 50488900
Encompass_Preferred
Irreversible serine protease inhibitor of chymotrypsin, kallikrein, papain, subtilisin, thrombin and trypsin. Working Conentration: 0.1-1.0mM.
Catalog No. 50-488-900 Supplier Research Products International Corp Supplier No. P2027025.0
Irreversible serine protease inhibitor of chymotrypsin, kallikrein, papain, subtilisin, thrombin and trypsin. Working Conentration: 0.1-1.0mM.