Learn More
Research Products International Corp PMSF, 250 G
SDP
Catalog No. 501976896
Encompass_Preferred
Irreversible serine protease inhibitor of chymotrypsin, kallikrein, papain, subtilisin, thrombin and trypsin. Working Conentration: 0.1-1.0mM.
Catalog No. 50-197-6896 Supplier Research Products International Corp Supplier No. P20270250.0
Add to Cart
Add to Cart
Irreversible serine protease inhibitor of chymotrypsin, kallikrein, papain, subtilisin, thrombin and trypsin. Working Conentration: 0.1-1.0mM.