Learn More
Bachem Phenylmethylsulfonyl fluoride
SDP
Catalog No. 50260623
Encompass_Preferred
Phenylmethylsulfonyl fluoride, 329-98-6, C7H7FO2S, Mr 174.20, 25g. PMSF, Pms-F. PMSF inhibits serine proteases such as trypsin and chymotrypsin, cysteine proteases such as papain, and acetylcholinesterase.
Catalog No. 50-260-623 Supplier Bachem Supplier No. 4000472.0025
Phenylmethylsulfonyl fluoride, 329-98-6, C7H7FO2S, Mr 174.20, 25g. PMSF, Pms-F. PMSF inhibits serine proteases such as trypsin and chymotrypsin, cysteine proteases such as papain, and acetylcholinesterase.