Learn More
Ubpbio PMSF, 25G
Supplier:  Ubpbio P105025
Encompass_Preferred
PMSF (phenylmethylsulfonyl Fluoride) is a serine protease inhibitor that reacts with the catalytic serine residue of trypsin, chymotrypsin, thrombin and papain at the concentration of 0.1-1 mM. PMSF has a relatively short life time in aqueous solutions (~ 50 min), it should be added in each protein purification step.
Catalog No. 50-196-5090