Learn More
Medchemexpress LLC HY-B0496 500mg , PMSF Phenylmethylsulfonyl fluoride;Benzylsulfonyl fluoride CAS:329-98-6 Purity:98%
SDP
Catalog No. 501492495
Encompass_Preferred
Medchemexpress, HY-B0496 500mg PMSF Phenylmethylsulfonyl fluoride;Benzylsulfonyl fluoride CAS:329-98-6 Formula:C7H7FO2S Purity:98% PMSF is an irreversible serine/cysteine protease inhibitor. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2495 Supplier Medchemexpress LLC Supplier No. HYB0496500MG
Medchemexpress, HY-B0496 500mg PMSF Phenylmethylsulfonyl fluoride;Benzylsulfonyl fluoride CAS:329-98-6 Formula:C7H7FO2S Purity:98% PMSF is an irreversible serine/cysteine protease inhibitor. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.