Learn More
Selleck Chemical LLC PMSF S3025-1g
SDP
Catalog No. 503111323
Encompass_Preferred
PMSF (Phenylmethylsulfonyl Fluoride Benzylsulfonyl fluoride) is an irreversible serine/cysteine protease inhibitor
Catalog No. 50-311-1323 Supplier Selleck Chemical LLC Supplier No. S30251G
PMSF (Phenylmethylsulfonyl Fluoride Benzylsulfonyl fluoride) is an irreversible serine/cysteine protease inhibitor