Learn More
Selleck Chemical LLC PMSF 50mg 329-98-6 Phenylmethylsulfonyl Fluoride
SDP
Catalog No. 501363273
Encompass_Preferred
PMSF (Phenylmethylsulfonyl Fluoride) is an irreversible serine/cysteine protease inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3273 Supplier Selleck Chemical LLC Supplier No. S302550MG
PMSF (Phenylmethylsulfonyl Fluoride) is an irreversible serine/cysteine protease inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.