Learn More
Ubpbio PMSF, 5G
Catalog No. 501126943
Encompass_Preferred
Protein Purification Reagents – PMSF (UBPBio catalog# P1050-5, P1050-25, P1050-100) – PMSF (phenylmethylsulfonyl Fluoride) is a serine protease inhibitor that reacts with the catalytic serine residue of trypsin, chymotrypsin, thrombin and papain at the concentration of 0.1-1 mM. PMSF has a relatively short life time in aqueous solutions (~ 50 min), it should be added in each protein purification step.
Catalog No. 50-112-6943 Supplier Ubpbio Supplier No. P10505
Protein Purification Reagents – PMSF (UBPBio catalog# P1050-5, P1050-25, P1050-100) – PMSF (phenylmethylsulfonyl Fluoride) is a serine protease inhibitor that reacts with the catalytic serine residue of trypsin, chymotrypsin, thrombin and papain at the concentration of 0.1-1 mM. PMSF has a relatively short life time in aqueous solutions (~ 50 min), it should be added in each protein purification step.