Learn More
Medchemexpress LLC PMSF (Phenylmethylsulfonyl fluoride) | 329-98-6 | 99.3% | 1 ML
SDP
Catalog No. 5000406724
Encompass_Preferred
PMSF (Phenylmethylsulfonyl fluoride) is a nonspecific and irreversible inhibitor for serine proteases. It also inhibits some thiol proteases, non-protease enzymes, and acetylcholinesterase, making it suitable for preparing protein extracts from tissue and cell lysates.
  • Nonspecific and irreversible serine proteases inhibitor
  • Inhibits certain thiol proteases, non-protease enzymes, and acetylcholinesterase
  • Used for preparing protein extracts
Catalog No. 50-004-06724 Supplier Medchemexpress LLC Supplier No. HYB049610MM/1ML
Add to Cart
Add to Cart
PMSF (Phenylmethylsulfonyl fluoride) is a nonspecific and irreversible inhibitor for serine proteases. It also inhibits some thiol proteases, non-protease enzymes, and acetylcholinesterase, making it suitable for preparing protein extracts from tissue and cell lysates.
  • Nonspecific and irreversible serine proteases inhibitor
  • Inhibits certain thiol proteases, non-protease enzymes, and acetylcholinesterase
  • Used for preparing protein extracts