Learn More
Cayman Chemical p-methyl-N-salicyloyl Tryptamine, Purity: ≥98%, CAS: 1371564-45-2, 10 mg
SDP
Catalog No. 502918888
Encompass_Preferred
A neuroprotective agent; reduces LPS-induced NO production in C6 cells and BV-2 microglia (IC50s = 7.48 and 4.94 µM, respectively); reduces LPS-induced production of TNF-α and PGE2 and increases production of IL-10 in BV-2 microglia at 40 µM; decreases cortical and hippocampal Aβ plaque formation and neuronal pyroptosis, as well as improves spatial learning and memory in a mouse model of Aβ-induced Alzheimer’s disease from 25-100 mg/kg
Catalog No. 50-291-8888 Supplier Cayman Chemical Supplier No. 3895810 MG
A neuroprotective agent; reduces LPS-induced NO production in C6 cells and BV-2 microglia (IC50s = 7.48 and 4.94 µM, respectively); reduces LPS-induced production of TNF-α and PGE2 and increases production of IL-10 in BV-2 microglia at 40 µM; decreases cortical and hippocampal Aβ plaque formation and neuronal pyroptosis, as well as improves spatial learning and memory in a mouse model of Aβ-induced Alzheimer’s disease from 25-100 mg/kg