Learn More
SAI Infusion Technologies 1.3mll Na Citrate Blood Tubes (3.2%) with Screw Cap
SDP
Catalog No. NC1942055
Encompass
1.3ml 3.2% Sodium Citrate Blood Tube with Screw Cap. Ideal for collection of 1.3ml of blood. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/box. Product is for laboratory animal research only.
Catalog No. NC1942055 Supplier SAI Infusion Technologies Supplier No. PMTSC1.3
1.3ml 3.2% Sodium Citrate Blood Tube with Screw Cap. Ideal for collection of 1.3ml of blood. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/box. Product is for laboratory animal research only.