Learn More
Medchemexpress LLC HY-110136A 1mg Medchemexpress, PMX 205 Trifluoroacetate CAS: Purity:>98%
SDP
Catalog No. 501968754
Encompass_Preferred
Medchemexpress, HY-110136A 1mg PMX 205 Trifluoroacetate CAS: PMX 205 Trifluoroacetate is a potent complement C5a receptor (C5aR; CD88) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8754 Supplier Medchemexpress LLC Supplier No. HY110136A1MG
Medchemexpress, HY-110136A 1mg PMX 205 Trifluoroacetate CAS: PMX 205 Trifluoroacetate is a potent complement C5a receptor (C5aR; CD88) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.