Learn More
Apexbio Technology LLC PMX 205 Trifluoroacetate 514814-49-4 (free base) 1mg
SDP
Catalog No. 5000092291
Encompass_Preferred
PMX 205 Trifluoroacetate is a cyclic hexapeptide antagonist targeting the complement C5a receptor (C5aR CD88) It is designed to competitively inhibit C5a binding to its receptor thereby modulating complement-mediated inflammatory pathways PMX 205 Trifluoroacetate exerts its biological activity primarily through competitive antagonism at C5aR reducing downstream inflammatory cellular responses In animal models PMX 205 Trifluoroacetate demonstrates anti-inflammatory effects Based on these pharmacological properties PMX 205 Trifluoroacetate holds research potential in the study of neuroinflammation Alzheimer s disease and chemically induced colitis
Catalog No. 50-000-92291 Supplier Apexbio Technology LLC Supplier No. B77871
PMX 205 Trifluoroacetate is a cyclic hexapeptide antagonist targeting the complement C5a receptor (C5aR CD88) It is designed to competitively inhibit C5a binding to its receptor thereby modulating complement-mediated inflammatory pathways PMX 205 Trifluoroacetate exerts its biological activity primarily through competitive antagonism at C5aR reducing downstream inflammatory cellular responses In animal models PMX 205 Trifluoroacetate demonstrates anti-inflammatory effects Based on these pharmacological properties PMX 205 Trifluoroacetate holds research potential in the study of neuroinflammation Alzheimer s disease and chemically induced colitis