Learn More
Medchemexpress LLC PMX 205 Trifluoroacetate | 99.5% | 953.06 | 100 MG
SDP
Catalog No. 5000344790
Encompass_Preferred
PMX 205 Trifluoroacetate is identified as a specific complement C5aR1 isoform antagonist. It is intended for research use only and is not sold to patients.
  • Specific complement C5aR1 isoform antagonist.
  • Orally active, selective C5aR antagonist.
  • Extends survival time and reduces end-stage motor scores in SOD1G93A rats.
  • Reduces astroglial proliferation in the lumbar spinal cord.
  • Decreases fibrillar plaque load in Tg2576 mice.
  • Significantly reduces hyperphosphorylated tau in 3xTg mice.
Catalog No. 50-003-44790 Supplier Medchemexpress LLC Supplier No. HY110136A100MG
PMX 205 Trifluoroacetate is identified as a specific complement C5aR1 isoform antagonist. It is intended for research use only and is not sold to patients.
  • Specific complement C5aR1 isoform antagonist.
  • Orally active, selective C5aR antagonist.
  • Extends survival time and reduces end-stage motor scores in SOD1G93A rats.
  • Reduces astroglial proliferation in the lumbar spinal cord.
  • Decreases fibrillar plaque load in Tg2576 mice.
  • Significantly reduces hyperphosphorylated tau in 3xTg mice.