Learn More
Medchemexpress LLC PMX 205 Trifluoroacetate | 99.5% | 953.06 | 100 MG
SDP
Catalog No. 5000345301
Encompass_Preferred
PMX 205 Trifluoroacetate is a specific complement C5aR1 isoform antagonist.
  • Specific complement C5aR1 isoform antagonist
  • Orally active
  • Extends survival time and reduces end-stage motor scores in a rat model of amyotrophic lateral sclerosis
  • Reduces astroglial proliferation in the lumbar spinal cord
  • Decreases fibrillar plaque load and reduces hyperphosphorylated tau in murine models of Alzheimer's disease
  • Available in various quantities and forms
Catalog No. 50-003-45301 Supplier Medchemexpress LLC Supplier No. HY11013610MM/1ML
PMX 205 Trifluoroacetate is a specific complement C5aR1 isoform antagonist.
  • Specific complement C5aR1 isoform antagonist
  • Orally active
  • Extends survival time and reduces end-stage motor scores in a rat model of amyotrophic lateral sclerosis
  • Reduces astroglial proliferation in the lumbar spinal cord
  • Decreases fibrillar plaque load and reduces hyperphosphorylated tau in murine models of Alzheimer's disease
  • Available in various quantities and forms