Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PMX 205 Trifluoroacetate | 99.50% | 953.06 | 1 ML
SDP

Catalog No. 5000344064
Encompass_Preferred

PMX 205 Trifluoroacetate is a specific complement C5aR1 isoform antagonist. It is intended for research use only and not for sale to patients.

  • Specific complement C5aR1 isoform antagonist.
  • Purity of 99.50%.
  • Molecular weight of 953.06.
  • Chemical formula is C47H63F3N10O8.
  • Appears as a white to off-white solid.
  • Soluble in DMSO at ≥ 100 mg/mL.
  • Shows significant inhibition of cell viability in MTT assays.
  • Functions as an orally active, selective C5aR antagonist in rats.
  • Reduces fibrillar plaque load in Tg2576 mice.
  • Significantly reduces hyperphosphorylated tau in 3xTg mice.

Catalog No. 50-003-44064 Supplier Medchemexpress LLC Supplier No. HY110136A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PMX 205 Trifluoroacetate is a specific complement C5aR1 isoform antagonist. It is intended for research use only and not for sale to patients.

  • Specific complement C5aR1 isoform antagonist.
  • Purity of 99.50%.
  • Molecular weight of 953.06.
  • Chemical formula is C47H63F3N10O8.
  • Appears as a white to off-white solid.
  • Soluble in DMSO at ≥ 100 mg/mL.
  • Shows significant inhibition of cell viability in MTT assays.
  • Functions as an orally active, selective C5aR antagonist in rats.
  • Reduces fibrillar plaque load in Tg2576 mice.
  • Significantly reduces hyperphosphorylated tau in 3xTg mice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.