Learn More
Medchemexpress LLC PMX-53 | 219639-75-5 | MFCD07363866 | 99.9% | 896.09 | C47H65N11O7 | 50 MG
SDP
Catalog No. 5000243819
Encompass_Preferred
PMX-53 is a synthetic peptidic antagonist of the complement C5a receptor (C5aR1/CD88). It is orally active and is used in preclinical research on inflammation, cancer, and atherosclerosis. Reported potency at C5aR1 is approximately 20 nM, and analytical data and storage recommendations are provided with the product documentation.
  • Peptidic C5a receptor antagonist.
  • Orally active, suitable for in vivo and in vitro studies.
  • Potent activity against C5aR1 (IC50 ≈ 20 nM).
  • Selective for C5aR1 over C5L2 and C3aR.
  • Characterized sequence and molecular weight for analytical use.
  • COA available with storage recommendations to preserve stability.
Catalog No. 50-002-43819 Supplier Medchemexpress LLC Supplier No. HY10617850MG
PMX-53 is a synthetic peptidic antagonist of the complement C5a receptor (C5aR1/CD88). It is orally active and is used in preclinical research on inflammation, cancer, and atherosclerosis. Reported potency at C5aR1 is approximately 20 nM, and analytical data and storage recommendations are provided with the product documentation.
  • Peptidic C5a receptor antagonist.
  • Orally active, suitable for in vivo and in vitro studies.
  • Potent activity against C5aR1 (IC50 ≈ 20 nM).
  • Selective for C5aR1 over C5L2 and C3aR.
  • Characterized sequence and molecular weight for analytical use.
  • COA available with storage recommendations to preserve stability.