Learn More
Selleck Chemical LLC PMX-53
SDP
Catalog No. 5000003877
Encompass_Preferred
PMX-53 is a potent CD88 (C5aR) antagonist and inhibits C5a-induced neutrophil myeloperoxidase release and chemotaxis with IC50 values of 22 nM and 75 nM respectively PMX-53 is also an agonist of Mas-related gene 2 (MrgX2)
Catalog No. 50-000-03877 Supplier Selleck Chemical LLC Supplier No. S623910MM/1ML
PMX-53 is a potent CD88 (C5aR) antagonist and inhibits C5a-induced neutrophil myeloperoxidase release and chemotaxis with IC50 values of 22 nM and 75 nM respectively PMX-53 is also an agonist of Mas-related gene 2 (MrgX2)