Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pmx-53 (Ac-Phe-{Orn}-Pro-{dCha}-Trp-Arg lactam) | 219639-75-5 | 99.9% | 896.09 g/mol | C47H65N11O7 | 100 MG
SDP

Catalog No. 5000243820
Encompass_Preferred

PMX-53 is a synthetic peptidic research compound that functions as a potent, orally active antagonist of the complement C5a receptor (C5aR1/CD88). It selectively binds C5aR1, displays low-affinity agonism at MrgX2, and is used in preclinical studies of inflammation, nociception, cancer, and atherosclerosis.

  • Potent complement C5a receptor antagonist (IC50 = 20 nM).
  • Inhibits C5a-induced neutrophil myeloperoxidase release (IC50 = 22 nM).
  • Inhibits C5a-induced neutrophil chemotaxis (IC50 = 75 nM).
  • Low-affinity MrgX2 agonist that can induce mast cell degranulation.
  • Peptidic sequence: Ac-Phe-{Orn}-Pro-{dCha}-Trp-Arg with Orn2-Arg6 lactam bridge.
  • Molecular formula C47H65N11O7; molecular weight 896.09 g/mol.
  • Available in small research pack sizes for in vitro and in vivo studies.

Catalog No. 50-002-43820 Supplier Medchemexpress LLC Supplier No. HY106178100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PMX-53 is a synthetic peptidic research compound that functions as a potent, orally active antagonist of the complement C5a receptor (C5aR1/CD88). It selectively binds C5aR1, displays low-affinity agonism at MrgX2, and is used in preclinical studies of inflammation, nociception, cancer, and atherosclerosis.

  • Potent complement C5a receptor antagonist (IC50 = 20 nM).
  • Inhibits C5a-induced neutrophil myeloperoxidase release (IC50 = 22 nM).
  • Inhibits C5a-induced neutrophil chemotaxis (IC50 = 75 nM).
  • Low-affinity MrgX2 agonist that can induce mast cell degranulation.
  • Peptidic sequence: Ac-Phe-{Orn}-Pro-{dCha}-Trp-Arg with Orn2-Arg6 lactam bridge.
  • Molecular formula C47H65N11O7; molecular weight 896.09 g/mol.
  • Available in small research pack sizes for in vitro and in vivo studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.