Learn More
Tocris Bioscience™ PMX 53

Description
Negative Control also available.
Specifications
Specifications
| Purity | 0.95 |
| Description | PMX 53 |
| Molecular Weight (g/mol) | 896.1 |
| Product Type | PMX 53 |
| Quantity | 1 mg |
| Sequence | Ac-Phe-cyclo(Orn-Pro-dCha-Trp-Arg) |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.