Learn More
Pyramex™ PMXSLIM™ Safety Eyewear
Catalog No. 19155812 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Bronze
Clear
Gray
Indoor/Outdoor, Mirror
Lens Coating:
UV-treated
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Coating
19-155-812 Clear -
19-155-813 Clear UV-treated
19-155-815 Gray UV-treated
19-155-816 Bronze -
19-155-814 Gray -
19-155-817 Indoor/Outdoor, Mirror -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-155-812 Supplier Pyramex™ Supplier No. SB7110S
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Slim fit for smaller facial features
  • Soft, adjustable PVC nose piece
  • Dual injected temple design includes soft rubber tips for a non-slip fit
  • Wrap-around, single lens design with incredible 10 base curve with side protection that extends to the temples
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Certifications/Compliance (Europe) EN-166
    Lens Tint Clear
    Frame Color Black
    Weight (Metric) 25 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate