Learn More
Pyramex™ PMXTREME™ Readers Safety Eyewear
Catalog No. 19155781 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Type:
Black frame with built-in swiveling LED bulbs
Black frame with cord
Magnification Power:
+1.50
+2.00
+2.50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Type Magnification Power
19-155-781 Black frame with cord +2.50
19-155-780 Black frame with cord +2.00
19-155-784 Black frame with built-in swiveling LED bulbs +1.50
19-155-785 Black frame with built-in swiveling LED bulbs +2.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-155-781 Supplier Pyramex™ Supplier No. SB6310SPR25
Add to Cart
Add to Cart
This item is not returnable. View return policy
Polycarbonate lens with nylon frame
  • Built-in rubber nose piece provides all day comfort
  • Includes black lanyard cord for extra security while wearing
  • Rubber temple tips keep eyewear secure for the active worker
  • Extreme style in safety eyewear, with clean lines and sports appeal
  • Ventilated, straight-back flexible temples provide cool, pressure-free fit
  • Scratch resistant polycarbonate lenses provide maximum vision protection and filter out 99% of harmful UV rays
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Tint Clear
    Magnification Power +2.50
    Weight (Metric) 27 g
    Lens Material Polycarbonate
    Frame Material Nylon
    Frame Type Black frame with cord