Learn More
Pyramex™ PMXTREME™ Safety Eyewear
Catalog No. 19155978 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
Camouflage
Red
Lens Tint:
Amber
Blue
Blue, Mirror
Bronze
Clear
Gray
Green, Mirror
Indoor/Outdoor, Mirror
Orange, Mirror
Silver, Mirror
Lens Coating:
Anti-fog, UV-treated
UV-treated
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
19-155-978 Red Gray -
19-155-777 Black Clear -
19-155-782 Black Clear UV-treated
19-155-788 Black Gray -
19-155-789 Black Gray Anti-fog, UV-treated
19-155-791 Black Amber -
19-155-792 Black Orange, Mirror -
19-155-793 Black Blue -
19-155-794 Black Silver, Mirror -
19-155-795 Black Blue, Mirror -
19-155-796 Black Green, Mirror -
19-155-797 Black Indoor/Outdoor, Mirror -
19-155-927 Camouflage Bronze UV-treated
19-155-928 Camouflage Gray UV-treated
19-155-926 Camouflage Clear Anti-fog, UV-treated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 19-155-978 Supplier Pyramex™ Supplier No. SR6320SP
Add to Cart
Add to Cart
This item is not returnable. View return policy
Offers protection from excessive glare
  • Built-in rubber nose piece provides all day comfort
  • Includes black lanyard cord for extra security while wearing
  • Rubber temple tips keep eyewear secure for the active worker
  • Extreme style in safety eyewear, with clean lines and sports appeal
  • Ventilated, straight-back flexible temples provide cool, pressure-free fit
  • Scratch resistant polycarbonate lenses provide maximum vision protection and filter out 99% of harmful UV rays
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Tint Gray
    Frame Color Red
    Weight (Metric) 27 g
    Lens Material Polycarbonate
    Frame Material Nylon