Learn More
Cayman Chemical pn-TrkC-Term RabbIt Monc 100uL
SDP
Catalog No. 502909329
Encompass_Preferred
A positive allosteric modulator of M1 mAChRs increases ACh-induced calcium flux in CHO cells expressing human M1 receptors (EC50 21 nM) reverses scopolamine-induced cognitive deficits in an object retrieval detour task in rhesus macaques at 0 3 and 1 mg/kg
Catalog No. 50-290-9329 Supplier Cayman Chemical Supplier No. 35316100 UL
A positive allosteric modulator of M1 mAChRs increases ACh-induced calcium flux in CHO cells expressing human M1 receptors (EC50 21 nM) reverses scopolamine-induced cognitive deficits in an object retrieval detour task in rhesus macaques at 0 3 and 1 mg/kg