Learn More
Vector Laboratories BPL, Unconjugated, 5 mg CL-1073-1
SDP
Supplier:  Vector Laboratories CL10731
Encompass
Bauhinia Purpurea Lectin (BPL), Unconjugated, 5 mg. BPL binding appears to be highest for glycoconjugates containing galactosyl (beta-1,3) N-acetylgalactosamine structures, but oligosaccharides with a terminal alpha-linked N-acetylgalactosamine can also bind. Although binding specificity is similar to that of peanut agglutinin, tissue staining patterns of these two lectins are distinct.
Catalog No. 50-204-5632