Learn More
Abcam pNA (p-nitroaniline), Solvatochromic dye, 100MG
Catalog No. 502705133
Encompass_Preferred
MW 138.12 Da, Purity >99%. Solvatochromic dye. UV-visual peak position changes with the balance of hydrogen bonding acceptors and donors in the solvent. Reacts to form a purple azo dye. Undergoes photocatalytic degradation in the presence of TiO₂ suspensions.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5133 Supplier Abcam Supplier No. AB145588100MG
MW 138.12 Da, Purity >99%. Solvatochromic dye. UV-visual peak position changes with the balance of hydrogen bonding acceptors and donors in the solvent. Reacts to form a purple azo dye. Undergoes photocatalytic degradation in the presence of TiO₂ suspensions.
The product is subject to the following: Abcam Restricted Use Statement