Learn More
Cayman Chemical pNA Stndard 50uL
SDP
Catalog No. 502944703
Encompass_Preferred
A riboside form of nicotinamide a precursor of NAD and a source of vitamin B3 increases intracellular and mitochondrial NAD content in C2C12 Hepa1.6 and HEK293 cells in a concentration-dependent manner at concentrations ranging from 1-1000 M decreases acetylation of FOXO1 and SOD2 but not tubulin increases NAD levels in liver and skeletal muscle and prevents body weight gain in mice fed a high-fat diet when used at a dose of 400 mg/kg per day increases NAD in the cerebral cortex and reduces cognitive deterioration in a transgenic mouse model of Alzheimers disease
Catalog No. 50-294-4703 Supplier Cayman Chemical Supplier No. 40051550 UL
A riboside form of nicotinamide a precursor of NAD and a source of vitamin B3 increases intracellular and mitochondrial NAD content in C2C12 Hepa1.6 and HEK293 cells in a concentration-dependent manner at concentrations ranging from 1-1000 M decreases acetylation of FOXO1 and SOD2 but not tubulin increases NAD levels in liver and skeletal muscle and prevents body weight gain in mice fed a high-fat diet when used at a dose of 400 mg/kg per day increases NAD in the cerebral cortex and reduces cognitive deterioration in a transgenic mouse model of Alzheimers disease