Learn More
Cayman Chemical Panaxadiol, Purity: ≥98%, CAS: 19666-76-3, 50 mg
SDP
Catalog No. 502946320
Encompass_Preferred
A triterpenoid sapogenin; inhibits the growth of HepG2, A549, MCF-7, and HCT116 cancer cells (IC50s = 63.20, 40.88, 61.96, and 92.53 µM, respectively); reduces tumor growth in an HCT116 mouse xenograft model at 30 mg/kg; inhibits HBV DNA replication in HepG 2.2.15 cells (IC50 = 148.15 µM); decreases the escape latency in the MWM in an APP/PS1 mouse model of Alzheimer’s disease at 50 mg/kg
Catalog No. 50-294-6320 Supplier Cayman Chemical Supplier No. 3323350 MG
A triterpenoid sapogenin; inhibits the growth of HepG2, A549, MCF-7, and HCT116 cancer cells (IC50s = 63.20, 40.88, 61.96, and 92.53 µM, respectively); reduces tumor growth in an HCT116 mouse xenograft model at 30 mg/kg; inhibits HBV DNA replication in HepG 2.2.15 cells (IC50 = 148.15 µM); decreases the escape latency in the MWM in an APP/PS1 mouse model of Alzheimer’s disease at 50 mg/kg