Learn More
Cayman Chemical Panaxatriol, Purity: ≥95%, CAS: 32791-84-7, 50 mg
SDP
Catalog No. 502940433
Encompass_Preferred
A sapogenin with diverse biological activities; inhibits the proliferation of human DU15 prostate cancer cells (IC50 = 30 μM) and induces apoptosis by increasing production of ROS and decreasing mitochondrial membrane potential; reduces ischemia and reperfusion-induced myocardial dysfunction, increases LVDevP and GSH levels, and decreases cardiac LDH, CK, and MDA levels in isolated rats at 5 mg/kg for seven consecutive days prior to injury; decreases fasting blood glucose levels in KKAy insulin-resistant mice at 0.2% in chow for 6 weeks
Catalog No. 50-294-0433 Supplier Cayman Chemical Supplier No. 2883550 MG
A sapogenin with diverse biological activities; inhibits the proliferation of human DU15 prostate cancer cells (IC50 = 30 μM) and induces apoptosis by increasing production of ROS and decreasing mitochondrial membrane potential; reduces ischemia and reperfusion-induced myocardial dysfunction, increases LVDevP and GSH levels, and decreases cardiac LDH, CK, and MDA levels in isolated rats at 5 mg/kg for seven consecutive days prior to injury; decreases fasting blood glucose levels in KKAy insulin-resistant mice at 0.2% in chow for 6 weeks