Learn More
Lonza Walkersville PNBM Primary Neuron Basal Med
Catalog No. 5000496440
Encompass
Clonetics PNBM Primary Neuron Basal Medium (CC-3256) specifically developed for culturing embryonic primary rat and mouse neurons For successful culture of neurons basal media should be combined with growth factors (sold seperately) Basal medium and SingleQuots Growth Factors can be purchased seperately or in convenient PNGM BulletKit format (CC-4461)
Catalog No. 50-004-96440 Supplier Lonza Walkersville Supplier No. CC3256
Clonetics PNBM Primary Neuron Basal Medium (CC-3256) specifically developed for culturing embryonic primary rat and mouse neurons For successful culture of neurons basal media should be combined with growth factors (sold seperately) Basal medium and SingleQuots Growth Factors can be purchased seperately or in convenient PNGM BulletKit format (CC-4461)