Learn More
Medchemexpress LLC PNC-27 acetate | 99.9% | 4031.73 | 50 MG
SDP
Catalog No. 5000399549
Encompass_Preferred
PNC-27 acetate is a chimeric p53-penetratin peptide that binds to HDM-2 in a p53 peptide-like structure. It induces selective membrane-pore formation and leads to cancer cell lysis. PNC-27 acetate is an anticancer peptide and can be used in acute myeloid leukemia research.
  • Induces cancer cell death.
  • Binds to cell membrane-bound HDM-2 in A2058 and MCF-7 cells.
  • Shows anti-leukemia activity in mice.
  • Reduces AML engraftment and prolongs survival in AML mice.
Catalog No. 50-003-99549 Supplier Medchemexpress LLC Supplier No. HYP3508A50MG
PNC-27 acetate is a chimeric p53-penetratin peptide that binds to HDM-2 in a p53 peptide-like structure. It induces selective membrane-pore formation and leads to cancer cell lysis. PNC-27 acetate is an anticancer peptide and can be used in acute myeloid leukemia research.
  • Induces cancer cell death.
  • Binds to cell membrane-bound HDM-2 in A2058 and MCF-7 cells.
  • Shows anti-leukemia activity in mice.
  • Reduces AML engraftment and prolongs survival in AML mice.