Learn More
Medchemexpress LLC PNC-27 acetate | 99.98% | 4031.73 (free base) | 10 MG
SDP
Catalog No. 5000397269
Encompass_Preferred
PNC-27 acetate is a chimeric p53-penetratin peptide that binds to HDM-2 in a p53 peptide-like structure, inducing selective membrane-pore formation and leading to cancer cell lysis. It is an anticancer peptide that can be used in acute myeloid leukemia research.
  • Binds to HDM-2 in a p53 peptide-like structure.
  • Induces selective membrane-pore formation.
  • Leads to cancer cell lysis.
  • Anticancer peptide.
  • Can be used in acute myeloid leukemia research.
  • Induces cancer cell death (e.g., 100% cell death in MIA-PaCa-2 cells in 90 minutes at 50 μg/mL).
  • Shows anti-leukemia activity in mice (e.g., reduced AML engraftment and prolonged survival at 40 mg/kg intraperitoneal injection once daily for 2-3 weeks).
Catalog No. 50-003-97269 Supplier Medchemexpress LLC Supplier No. HYP3508A10MG
PNC-27 acetate is a chimeric p53-penetratin peptide that binds to HDM-2 in a p53 peptide-like structure, inducing selective membrane-pore formation and leading to cancer cell lysis. It is an anticancer peptide that can be used in acute myeloid leukemia research.
  • Binds to HDM-2 in a p53 peptide-like structure.
  • Induces selective membrane-pore formation.
  • Leads to cancer cell lysis.
  • Anticancer peptide.
  • Can be used in acute myeloid leukemia research.
  • Induces cancer cell death (e.g., 100% cell death in MIA-PaCa-2 cells in 90 minutes at 50 μg/mL).
  • Shows anti-leukemia activity in mice (e.g., reduced AML engraftment and prolonged survival at 40 mg/kg intraperitoneal injection once daily for 2-3 weeks).