Learn More
Medchemexpress LLC PNC-27 acetate | 99.98% | 4031.73 (free base) | 100 MG
SDP
Catalog No. 5000396162
Encompass_Preferred
PNC-27 acetate is a chimeric p53-penetratin peptide that binds to HDM-2 in a p53 peptide-like structure.
  • Induces selective membrane-pore formation, leading to cancer cell lysis.
  • Utilized in acute myeloid leukemia research.
Catalog No. 50-003-96162 Supplier Medchemexpress LLC Supplier No. HYP3508A100MG
PNC-27 acetate is a chimeric p53-penetratin peptide that binds to HDM-2 in a p53 peptide-like structure.
  • Induces selective membrane-pore formation, leading to cancer cell lysis.
  • Utilized in acute myeloid leukemia research.