Learn More
Medchemexpress LLC PNC-28 acetate | 392661-17-5 | 98.8% | 3509.13 (free acid) | 10 MG
SDP
Catalog No. 5000399354
Encompass_Preferred
PNC-28 acetate is a peptide derived from the mdm-2-binding domain (residues 17-26) of the p53 protein, incorporating a membrane-crossing-penetratin sequence. This compound is suitable for use in pancreatic cancer research.
  • Can be used for pancreatic cancer research
  • Blocks growth of a lethal human pancreatic cancer cell line in vitro
  • Inhibits BMRPA1. Tuc3 tumor growth in vivo, particularly when delivered directly to the tumor
Catalog No. 50-003-99354 Supplier Medchemexpress LLC Supplier No. HYP3509A10MG
PNC-28 acetate is a peptide derived from the mdm-2-binding domain (residues 17-26) of the p53 protein, incorporating a membrane-crossing-penetratin sequence. This compound is suitable for use in pancreatic cancer research.
  • Can be used for pancreatic cancer research
  • Blocks growth of a lethal human pancreatic cancer cell line in vitro
  • Inhibits BMRPA1. Tuc3 tumor growth in vivo, particularly when delivered directly to the tumor