Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PNC-28 acetate | 98.8% | 3509.13 (free acid) | 1 MG
SDP

Catalog No. 5000397394
Encompass_Preferred

PNC-28 acetate is a peptide derived from the mdm-2-binding domain (residues 17-26) of the p53 protein, incorporating a membrane crossing-penetratin sequence. It is used for pancreatic cancer research. For research use only; not for patient use. PNC-28 blocks the growth of lethal human pancreatic cancer cell lines in vitro and inhibits tumor growth in vivo.

  • High purity of 98.79%
  • Suitable for in vitro and in vivo pancreatic cancer research
  • Supplied as a solid, white to off-white appearance
  • Sequence: Glu-Thr-Phe-Ser-Asp-Leu-Trp-Lys-Leu-Leu-Lys-Lys-Trp-Lys-Met-Arg-Rr-Asn-Gln-Phe-Trp-Val-Lys-Vl-Gln-Arg-Gly
  • Soluble in H2O at 100 mg/mL (requires ultrasonic)
  • Sealed storage under nitrogen, away from moisture and light is recommended

Catalog No. 50-003-97394 Supplier Medchemexpress LLC Supplier No. HYP3509A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PNC-28 acetate is a peptide derived from the mdm-2-binding domain (residues 17-26) of the p53 protein, incorporating a membrane crossing-penetratin sequence. It is used for pancreatic cancer research. For research use only; not for patient use. PNC-28 blocks the growth of lethal human pancreatic cancer cell lines in vitro and inhibits tumor growth in vivo.

  • High purity of 98.79%
  • Suitable for in vitro and in vivo pancreatic cancer research
  • Supplied as a solid, white to off-white appearance
  • Sequence: Glu-Thr-Phe-Ser-Asp-Leu-Trp-Lys-Leu-Leu-Lys-Lys-Trp-Lys-Met-Arg-Rr-Asn-Gln-Phe-Trp-Val-Lys-Vl-Gln-Arg-Gly
  • Soluble in H2O at 100 mg/mL (requires ultrasonic)
  • Sealed storage under nitrogen, away from moisture and light is recommended

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.