Learn More
Aviva Systems Biology Corp Pnck Antibody - C-terminal region (ARP51857_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51857_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pnck. It was validated on Western Blot. . Alternate names: Camk1bApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-603