Learn More
Biomatik Corporation Pnck Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11560
Encompass_Preferred
Pnck Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PNCK. The Pnck Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Calcium; Calmodulin-Dependent Protein Kinase Type 1B Protein (1-115Aa). Applications: ELISA, WB, IHC. Target Synonyms: PNCK. Accession Number: Q6P2M8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9240